missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Hydroxytetrahydrofuran, 98%

Product Code. 15492958
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15492958

Brand: Thermo Scientific Alfa Aesar H56476.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86087-23-2 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86087-23-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00064327
InChI Key XDPCNPCKDGQBAN-BYPYZUCNSA-N
Synonym s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s
PubChem CID 2733227
IUPAC Name (3S)-oxolan-3-ol
SMILES C1COCC1O

Specifications

Density 1.103
Boiling Point 80°C (15 mmHg)
Quantity 25 g
Refractive Index 1.45
Beilstein 4652609
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-3-Hydroxytetrahydrofuran

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.