missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Hydroxy-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-Dioxide, TRC

Product Code. 30391057
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391057 5 g 5g
30391327 25 g 25g
30402236 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391057 Supplier Toronto Research Chemicals Supplier No. TRCH9462005G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154127-42-1
Molecular Formula C10 H16 N2 O6 S3
Molecular Weight (g/mol) 356.44
Synonym (4S)-3,4-Dihydro-4-hydroxy-2-(3-methoxypropyl)-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, (S)-3,4-Dihydro-4-hydroxy-2-(3-methoxypropyl)-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide
IUPAC Name (4S)-4-hydroxy-2-(3-methoxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide
SMILES COCCCN1C[C@@H](O)c2cc(sc2S1(=O)=O)S(=O)(=O)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H16N2O6S3/c1-18-4-2-3-12-6-8(13)7-5-9(20(11,14)15)19-10(7)21(12,16)17/h5,8,13H,2-4,6H2,1H3,(H2,11,14,15)/t8-/m1/s1
Formula Weight 356.017
Chemical Name or Material (4S)-3,4-Dihydro-4-hydroxy-2-(3-methoxypropyl)-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.