missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Hydroxy Mephenytoin, TRC

Product Code. 30008382
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008382 5 mg 5mg
30003364 10 mg 10mg
30011061 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008382 Supplier Toronto Research Chemicals Supplier No. TRCH9448705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82695-93-0
Molecular Formula C12 H14 N2 O3
Molecular Weight (g/mol) 234.25
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-(4-hydroxyphenyl)-3-methyl-, (5S)-, 2,4-Imidazolidinedione, 5-ethyl-5-(4-hydroxyphenyl)-3-methyl-, (S)-, (5S)-5-Ethyl-5-(4-hydroxyphenyl)-3-methyl-2,4-imidazolidinedione, (S)-4-Hydroxymephenytoin, (S)-4'-Hydroxymephenytoin, (S)-4-Hydroxy mephenytoin
IUPAC Name (5S)-5-ethyl-5-(4-hydroxyphenyl)-3-methylimidazolidine-2,4-dione
SMILES CC[C@]1(NC(=O)N(C)C1=O)c2ccc(O)cc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H14N2O3/c1-3-12(8-4-6-9(15)7-5-8)10(16)14(2)11(17)13-12/h4-7,15H,3H2,1-2H3,(H,13,17)/t12-/m0/s1
Formula Weight 234.1004
Percent Purity >95%
Chemical Name or Material (S)-4-Hydroxy Mephenytoin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.