missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Hydroxymethyl Ambrisentan, TRC

Product Code. 30405326
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405326 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405326 Supplier Toronto Research Chemicals Supplier No. TRCH9477601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-4-Hydroxymethyl Ambrisentan
CAS 1106685-66-8
Molecular Formula C22H22N2O5
Quantity 1 mg
Synonym (alphaS)-alpha-[[4-(Hydroxymethyl)-6-methyl-2-pyrimidinyl]oxy]-beta-methoxy-beta-phenylbenzenepropanoic Acid
InChI Formula InChI=1S/C22H22N2O5/c1-15-13-18(14-25)24-21(23-15)29-19(20(26)27)22(28-2,16-9-5-3-6-10-16)17-11-7-4-8-12-17/h3-13,19,25H,14H2,1-2H3,(H,26,27)/t19-/m1/s1
SMILES COC([C@H](Oc1nc(C)cc(CO)n1)C(=O)O)(c2ccccc2)c3ccccc3
IUPAC Name (2S)-2-[4-(hydroxymethyl)-6-methylpyrimidin-2-yl]oxy-3-methoxy-3,3-diphenylpropanoic acid
Molecular Weight (g/mol) 394.42
Formula Weight 394.1529
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.