missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Methyl-2-pentanol, 99%, Thermo Scientific™

Product Code. 10741515 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10741515 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10741515 Supplier Thermo Scientific Acros Supplier No. 389230010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 14898-80-7
CAS Min % 98.5
CAS Max % 100.0
Density 0.8020g/mL
Boiling Point 132.0°C
Flash Point 41°C
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C6H14O
Refractive Index 1.4100 to 1.4120
MDL Number MFCD03093078
Quantity 1g
Specific Gravity 0.802
Specific Rotation Condition +20° (20°C c=1,CHCl3)
Specific Rotation + 20.00
Synonym s-+-4-methyl-2-pentanol, 2s-4-methylpentan-2-ol, s-+-4-methylpentan-2-ol, 4-methyl s-2-pentanol, s-4-methyl-pentan-2-ol, 2s-4-methyl-2-pentanol, s-1,3-dimethyl-1-butanol, 2-pentanol,4-methyl-, 2s, 2-pentanol, 4-methyl-, 2s, unii-8u34xjk0r0 component
InChI Key WVYWICLMDOOCFB-LURJTMIESA-N
SMILES CC(C)CC(C)O
IUPAC Name (2S)-4-methylpentan-2-ol
Molecular Weight (g/mol) 102.177
PubChem CID 6994471
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material (S)-(+)-4-Methyl-2-pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-4-Methyl-2-pentanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.