missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Methyl-2-pentanol, 99%

Product Code. 11369535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369535 250 mg 250mg
11359535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369535 Supplier Thermo Scientific Alfa Aesar Supplier No. L18882.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14898-80-7 | C6H14O | 102.177 g/mol

It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-80-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD03093078
InChI Key WVYWICLMDOOCFB-LURJTMIESA-N
PubChem CID 6994471
IUPAC Name (2S)-4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

Density 0.802
Boiling Point 130°C to 131°C
Flash Point 41°C (105°F)
Odor Slightly Sweetish
Refractive Index 1.411
Quantity 250 mg
UN Number UN2053
Beilstein 1718991
Synonym s-+-4-methyl-2-pentanol, 2s-4-methylpentan-2-ol, s-+-4-methylpentan-2-ol, 4-methyl s-2-pentanol, s-4-methyl-pentan-2-ol, 2s-4-methyl-2-pentanol, s-1,3-dimethyl-1-butanol, 2-pentanol,4-methyl-, 2s, 2-pentanol, 4-methyl-, 2s, unii-8u34xjk0r0 component
Solubility Information Slightly soluble in water. Soluble in alcohol.
Optical Rotation +21° (Neat)
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material (S)-(+)-4-Methyl-2-pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-4-Methyl-2-pentanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Supplemental information
  • MIXTURE LIST-Contain: (S)-(+)-4-Methyl-2-pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.