missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Phenyl-2-oxazolidinone, 98%, Thermo Scientific™

Product Code. 11352379
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352379 1 g 1g
11362379 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352379 Supplier Thermo Scientific Alfa Aesar Supplier No. H27306.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 99395-88-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00043396
InChI Key QDMNNMIOWVJVLY-MRVPVSSYSA-N
Synonym s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
PubChem CID 730424
IUPAC Name (4S)-4-phenyl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C2=CC=CC=C2

Especificaciones

Melting Point 130°C to 132°C
Quantity 1 g
Formula Weight 163.18
Optical Rotation +52° (c=2 in Chloroform)
Percent Purity 98%
Chemical Name or Material (S)-(+)-4-Phenyl-2-oxazolidinone
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.