missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Phenyl-2-oxazolidinone, 99%

Product Code. 10225781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225781 1 g Glass Bottle 1g
10663862 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225781 Supplier Thermo Scientific Acros Supplier No. 303850010
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 99395-88-7 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99395-88-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00043396
InChI Key QDMNNMIOWVJVLY-MRVPVSSYSA-N
Synonym s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076
PubChem CID 730424
IUPAC Name (4S)-4-phenyl-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)C2=CC=CC=C2

Specifications

Melting Point 130°C to 133°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Specific Rotation + 49.5
Specific Rotation Condition +49.5° (20°C c=2,CHCl3)
Quantity 1 g
Formula Weight 163.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-4-Phenyl-2-oxazolidinone, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.