missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-tert-Butyloxazolidine-2,5-dione, 98%, Thermo Scientific™

Product Code. 15409139
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15409139 Supplier Thermo Scientific Alfa Aesar Supplier No. L20222.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away fr
TRUSTED_SUSTAINABILITY

Specifications

CAS 62965-56-4
Melting Point 126°C to 128°C
Molecular Formula C7H11NO3
MDL Number MFCD05664067
Quantity 1 g
Beilstein 4664497
Sensitivity Moisture sensitive
Synonym s---4-tert-butyloxazolidine-2,5-dione, s-4-tert-butyloxazolidine-2,5-dione, s-4-tert-butyl oxazolidine-2,5-dione, 4s-4-tert-butyl-1,3-oxazolidine-2,5-dione, 4beta-tert-butyloxazolidine-2,5-dione, 2,5-oxazolidinedione,4-1,1-dimethylethyl-, 4s
Solubility Information Slightly soluble in water.
InChI Key GRDXOXRGSKKLNH-SCSAIBSYSA-N
Optical Rotation −41°
SMILES CC(C)(C)C1C(=O)OC(=O)N1
IUPAC Name (4S)-4-tert-butyl-1,3-oxazolidine-2,5-dione
Molecular Weight (g/mol) 157.169
PubChem CID 57351753
Formula Weight 157.17
Percent Purity 98%
Chemical Name or Material (S)-(-)-4-tert-Butyloxazolidine-2,5-dione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.