missing translation for 'onlineSavingsMsg'
Learn More

S-(5'-Adenosyl)-L-homocysteine, TRC

Product Code. 17932755
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932755 10 mg 10mg
17911975 100 mg 100mg
17900525 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932755 Supplier Toronto Research Chemicals Supplier No. TRCA29150010MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 979-92-0
Molecular Formula C14 H20 N6 O5 S
Synonym S-(5'-Deoxyadenosin-5'-yl)-L-homocysteine∣L-5'-S-(3-Amino-3-carboxypropyl)-5'-thioadenosine∣Adenosyl-L-homocysteine∣Adenosylhomocysteine∣L-S-Adenosylhomocysteine∣S-Adenosyl-L-homocysteine∣S-Adenosylhomocysteine;

Specifications

Format Neat
Molecular Weight (g/mol) 384.41
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 10 mg
Chemical Name or Material S-Adenosyl-L-Homocysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.