missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (S)-(-)-5-Iodowillardiine

Product Code. 30001459
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001459 10 mg 10mg
18066032 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001459 Supplier R&D Systems™ Supplier No. 0307/10
  • ONLINE EXCLUSIVE PRICE
    244.53 €
    213.15 € / 10mg
    Save 31.38 €
    12% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent and subtype-selective kainate agonist

Demonstrates high affinity for the kainate receptor subtype hGluK1 (formerly hGluR5) (Ki = 0.24 nM) and 600-4000-fold selectivity over both the AMPA receptor subtypes and the homomeric kainate receptor hGluK2 (formerly hGluR6).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140187-25-3
Molecular Formula C7H8IN3O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 325.06
Purity >98%
Molecular Weight (g/mol) 325.062
Chemical Name or Material (S)-(-)-5-Iodowillardiine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.