missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress S-Allyl-L-cysteine

Product Code. 16425457
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16425457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16425457 Supplier MedChemExpress Supplier No. HYW013573
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress S-Allyl-L-cysteine, one of the organosulfur compounds found in AGE, possess various biological effects including neurotrophic activity, anti-cancer activity, anti-inflammatory activity.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 161.22 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 21593-77-1
Product Type Natural Products
Purity 97.0%
Solubility H2O : 16.67 mg/mL (103.40 mM∣ultrasonic and warming and heat to 60°C)
Target Apoptosis
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.