missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-(1-Naphthyl)ethylamine, 99+%

Product Code. 10082900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10082900 1 g Glass bottle 1g
10299723 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082900 Supplier Thermo Scientific Acros Supplier No. 217810010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10420-89-0 | C12H14N | 172.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10420-89-0
Molecular Formula C12H14N
Molecular Weight (g/mol) 172.25
MDL Number MFCD00064179
InChI Key RTCUCQWIICFPOD-VIFPVBQESA-O
Synonym s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
PubChem CID 66325
IUPAC Name (1S)-1-naphthalen-1-ylethanamine
SMILES C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Orange to Yellow
Density 1.0600g/mL
Boiling Point 153.0°C (11.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH(CH3)NH2
Refractive Index 1.6217 to 1.6237
Quantity 1 g
Beilstein 12, III, 3111
Specific Gravity 1.06
Specific Rotation Condition - 60.00 (20.00°C c=2,CH3OH)
Specific Rotation − 60.00
Formula Weight 171.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (S)-(-)-α-(1-Naphthyl)ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-(1-Naphthyl)ethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.