missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-alpha-(Trifluoromethyl)benzyl alcohol, 99%

Product Code. 10765642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10765642 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10765642 Supplier Thermo Scientific Acros Supplier No. 428180010
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 340-06-7 | C8H7F3O | 176.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 340-06-7
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 127°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H7F3O
Linear Formula C6H5CH(OH)CF3
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
MDL Number MFCD00077845
Quantity 1 g
Specific Gravity 1.3
Specific Rotation +30° to +33° (20°C, 589nm) (C= neat)
Synonym s-+-alpha-trifluoromethyl benzyl alcohol, s-2,2,2-trifluoro-1-phenylethanol, 1s-2,2,2-trifluoro-1-phenylethan-1-ol, 1s-2,2,2-trifluoro-1-phenylethanol, s-1-phenyl-2,2,2-trifluoroethanol, s-+-1-phenyl-2,2,2-trifluoroethanol, pubchem15591, s-1-phenyltrifluoroethanol, s-a-trifluoromethyl benzyl alcohol, 1s-1-phenyl-2,2,2-trifluoroethanol
InChI Key VNOMEAQPOMDWSR-ZETCQYMHSA-N
SMILES C1=CC=C(C=C1)C(C(F)(F)F)O
IUPAC Name (1S)-2,2,2-trifluoro-1-phenylethanol
Molecular Weight (g/mol) 176.14
PubChem CID 1472401
Formula Weight 176.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(+)-α-(Trifluoromethyl)benzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-alpha-(Trifluoromethyl)benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.