missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (S)-AMPA

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
Active enantiomer of AMPA (EC50 = 3.5 μM).
NPEC-caged-(S)-AMPA, Inactive Enantiomer and Racemate also available.
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 186.17 |
| Purity | >99% |
| Molecular Weight (g/mol) | 186.167 |
| Chemical Name or Material | (S)-AMPA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction