missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (S)-(-)-Blebbistatin
GREENER_CHOICE

Product Code. 30001050
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001050 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001050 Supplier Tocris Bioscience™ Supplier No. 1852/1
  • ONLINE EXCLUSIVE PRICE
    300.00 €
    283.50 € / 1mg
    Save 16.50 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Selective inhibitor of myosin II ATPase activity. Active enantiomer of (±)-blebbistatin (Cat. No. 1760)

(S)-(-)-Blebbistatin is a selective inhibitor of myosin II ATPase activity (IC50 ∼0.5 - 5 μM); active enantiomer of (±)-blebbistatin. Inhibits contraction of the cleavage furrow without disrupting mitosis or contractile ring assembly. (S)-(-)-Blebbistatin rapidly and reversibly blocks cell blebbing, and disrupts directed cell migration and cytokinesis in vertebrate cells.

Inactive Enantiomer and Racemate also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 856925-71-8
Quantity 1 mg
Molecular Formula C18H16N2O2
Formula Weight 292.34
Purity >99%
Molecular Weight (g/mol) 292.34
Chemical Name or Material (S)-(-)-Blebbistatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.