missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Blebbistatin, TRC

Product Code. 30007115
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007115 50 mg 50mg
30012456 10 mg 10mg
30005111 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007115 Supplier Toronto Research Chemicals Supplier No. TRCB59250050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 856925-71-8
Molecular Formula C18 H16 N2 O2
Molecular Weight (g/mol) 292.33
Synonym 4H-Pyrrolo[2,3-b]quinolin-4-one, 1,2,3,3a-tetrahydro-3a-hydroxy-6-methyl-1-phenyl-, (3aS)-, (3aS)-1,2,3,3a-Tetrahydro-3a-hydroxy-6-methyl-1-phenyl-4H-pyrrolo[2,3-b]quinolin-4-one, (-)-Blebbistatin, (S)-(-)-Blebbistatin
IUPAC Name (3aS)-3a-hydroxy-6-methyl-1-phenyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-one
SMILES Cc1ccc2N=C3N(CC[C@@]3(O)C(=O)c2c1)c4ccccc4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H16N2O2/c1-12-7-8-15-14(11-12)16(21)18(22)9-10-20(17(18)19-15)13-5-3-2-4-6-13/h2-8,11,22H,9-10H2,1H3/t18-/m1/s1
Formula Weight 292.1212
Percent Purity >95%
Chemical Name or Material (S)-(-)-Blebbistatin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.