missing translation for 'onlineSavingsMsg'
Learn More

S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%

Product Code. 11450577
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450577 25 g 25g
11440577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450577 Supplier Thermo Scientific Alfa Aesar Supplier No. B22435.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41840-28-2 | C11H16N2O2S | 240.32 g/mol

S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41840-28-2
Molecular Formula C11H16N2O2S
Molecular Weight (g/mol) 240.32
MDL Number MFCD00006080
InChI Key POTDIELOEHTPJN-UHFFFAOYSA-N
Synonym s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester
PubChem CID 148428
IUPAC Name tert-butyl (4,6-dimethylpyrimidin-2-yl)sulfanylformate
SMILES CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1

Specifications

Melting Point 48°C to 51°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 396984
Formula Weight 240.33
Percent Purity 97%
Chemical Name or Material S-Boc-2-mercapto-4,6-dimethylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.