Learn More
Heidolph Instruments S-Box Package 1
Product Code. 17878000 Shop All Heidolph Instruments Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Violet
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17878000 Violet Each
17848000 Green Each
17858000 Orange Each
17868000 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17878000 Supplier Heidolph Instruments Supplier No. 4000500300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
The S-Box Package 1 includes additional accessories to complete an existing S-Biosystem incubation system.
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
TRUSTED_SUSTAINABILITY
Specifikationer
Color Violet
Includes 1 S-box, 1 Tool Kit, 4 inner boxes, 4 water bags, 4 front covers
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.