missing translation for 'onlineSavingsMsg'
Learn More

(S)-Camphanic Acid, TRC

Product Code. 30339805
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339805 Supplier Toronto Research Chemicals Supplier No. TRCC1751101G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material (S)-Camphanic Acid
CAS 13429-83-9
Molecular Formula C10H14O4
Quantity 1 g
Synonym Camphanic Acid, (-)-ω-Camphanic Acid, (-)-Camphanic Acid, (-)-ω-Camphanic Acid, (1S)-(-)-Camphanic Acid, (1S)-Camphanic Acid, (1S,4R)-(-)-Camphanic Acid, (1S,4R)-ω-Camphanic Acid, (1S)-4,7,7-Trimethyl-3-oxo-2-Oxabicyclo[2.2.1]heptane-1-carboxylic Acid
InChI Formula InChI=1S/C10H14O4/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3,(H,11,12)/t9-,10+/m0/s1
SMILES CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(=O)O
IUPAC Name (1R,4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carboxylic acid
Molecular Weight (g/mol) 198.22
Formula Weight 198.0892
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.