missing translation for 'onlineSavingsMsg'
Learn More

S-Carboxymethyl-L-cysteine, 98%

Product Code. 10187883 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187883 1 g Glass bottle 1g
10483965 5 g Glass bottle 5g
10656222 25 g Glass bottle 25g
10666222 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10187883 Supplier Thermo Scientific Supplier No. 225810010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-23-3 | C5H9NO4S | 179.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-23-3
Molecular Formula C5H9NO4S
Molecular Weight (g/mol) 179.19
MDL Number MFCD00002614
InChI Key GBFLZEXEOZUWRN-VKHMYHEASA-N
Synonym s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab
PubChem CID 193653
ChEBI CHEBI:16163
IUPAC Name (2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid
SMILES N[C@@H](CSCC(O)=O)C(O)=O

Specifications

Melting Point 205.0°C to 207.0°C
Color White
Ignition Residue 0.2% max.
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2SCH2CH(NH2)CO2H
Quantity 1 g
Merck Index 15, 1802
Specific Rotation Condition −31° (20°C c=1,10% NaH2CO3)
Specific Rotation − 31.00
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in hot water (3 g/100 mL), acids and alkal, slightly soluble in methanol and acetone
Formula Weight 179.19
Percent Purity 98%
Physical Form Amorphous Powder
Chemical Name or Material S-Carboxymethyl-L-cysteine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • S-Carboxymethyl-L-cysteine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.