missing translation for 'onlineSavingsMsg'
Learn More

(S)-cis,trans-Abscisic Acid Glucosyl Ester, TRC

Product Code. 30011134
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011134 10 mg 10mg
30003858 1 mg 1mg
30009023 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011134 Supplier Toronto Research Chemicals Supplier No. TRCA11012510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21414-42-6
Molecular Formula C21H30O9
Molecular Weight (g/mol) 426.46
Synonym beta-D-1-[(Z,E)-(+)-5-(1-hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-2,4-pentadienoate]glucopyranose, (+)-Abscisic acid glucose ester, (+)-Abscisyl-beta-D-glucopyranoside
IUPAC Name [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] (2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoate
SMILES C\C(=C\C(=O)O[C@H]1OC(CO)[C@@H](O)[C@H](O)[C@@H]1O)\C=C\[C@@]2(O)C(=CC(=O)CC2(C)C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H30O9/c1-11(5-6-21(28)12(2)8-13(23)9-20(21,3)4)7-15(24)30-19-18(27)17(26)16(25)14(10-22)29-19/h5-8,14,16-19,22,25-28H,9-10H2,1-4H3/b6-5+,11-7-/t14-,16-,17+,18-,19+,21-/m1/s1
Formula Weight 426.189
Percent Purity >95%
Chemical Name or Material (S)-cis,trans-Abscisic Acid Glucosyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.