missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ S-CLASSIC-90 2-Door Yellow Safety Storage Cabinets

Product Code. 15638845 Shop All asecos Products
Change view
Click to view available options
Configuration:
3 150 kg Tray Shelves, 1 135 kg Insert, 1 55 L Sump
4 x 60 kg/29 L Drawers
6 x 60 kg/19 L Drawers
Load Bearing Capacity:
3 x Tray shelves 150 kg/33 L each; Insert 135 kg; Sump 55 L
4 x Drawers 60 kg/29 L each
6 x Drawers 60 kg/19 L each
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Load Bearing Capacity Unit Size
15638845 4 x 60 kg/29 L Drawers 4 x Drawers 60 kg/29 L each Each
15769428 6 x 60 kg/19 L Drawers 6 x Drawers 60 kg/19 L each Each
15879216 3 150 kg Tray Shelves, 1 135 kg Insert, 1 55 L Sump 3 x Tray shelves 150 kg/33 L each; Insert 135 kg; Sump 55 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15638845 Supplier Asecos™ Supplier No. 3011600233724
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For the safe and approved storage of flammable hazardous substances in working areas, with an optional door open arrest system.

  • Aseco’s first EN-compliant safety storage cabinet
  • Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
  • Available in a range of widths and heights
  • Versatile fields of application due to various interior equipment
  • Convenient handling – door operation with minimal effort
  • Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
  • Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
  • Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • Fire resistance of 90 minutes
  • Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • In accordance with EN 14470-1 and TRGS 510

Specifications

Certifications/Compliance EN 14470-1
Configuration 4 x 60 kg/29 L Drawers
Dimensions (D x W x H) 616 x 1196 x 1968 mm
Door Style Triple-hinged
For Use With (Application) Storage of flammable and hazardous substances
Includes Door open arrest system
Load Bearing Capacity 4 x Drawers 60 kg/29 L each
No. of Doors 2
Surface Coating Powder-coated
Weight (Metric) 482 kg
Color Yellow
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.