missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-CLASSIC-90 One-Door Yellow Safety Storage Cabinets with Door Open Arrest System
Shop All asecos ProductsDescrizione
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifica
Specifica
| Certifications/Compliance | EN 14470-1, TRGS 510 |
| Configuration | 4 Drawers |
| Dimensions (D x W x H) | 616 x 596 x 1968 mm |
| Door Style | Triple-hinged |
| For Use With (Application) | Storage of flammable hazardous materials |
| Includes | Right-hinged door open arrest system |
| Load Bearing Capacity | 25 kg/5 L Drawers, Max Load 600 kg |
| No. of Doors | 1 |
| Surface Coating | Powder-coated |
| Weight (Metric) | 291 kg |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?