missing translation for 'onlineSavingsMsg'
Learn More

(S)-Efavirenz, TRC

Product Code. 30010097
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010097 10 mg 10mg
30009076 5 mg 5mg
30012785 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010097 Supplier Toronto Research Chemicals Supplier No. TRCE42500010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154598-52-4
Molecular Formula C14 H9 Cl F3 N O2
Molecular Weight (g/mol) 315.68
Synonym (4S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, (4S)-6-chloro-4-(cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, (S)-6-chloro-4-(cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, (-)-Efavirenz, (S)-4-(2-Cyclopropylethynyl)-6-chloro-4-(trifluoromethyl)-1,4-dihydro-2H-3,1-benzoxazin-2-one, (S)-4-(Cyclopropylethynyl)-6-chloro-4-(trifluoromethyl)-1,4-dihydro-2H-3,1-benzoxazin-2-one, (S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, DMP 266, EFV, Efavirenz, L 743726, Stocrin, Sustiva
IUPAC Name (4S)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one
SMILES FC(F)(F)[C@]1(OC(=O)Nc2ccc(Cl)cc12)C#CC3CC3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H9ClF3NO2/c15-9-3-4-11-10(7-9)13(14(16,17)18,21-12(20)19-11)6-5-8-1-2-8/h3-4,7-8H,1-2H2,(H,19,20)/t13-/m0/s1
Formula Weight 315.0274
Percent Purity >95%
Chemical Name or Material Efavirenz
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.