missing translation for 'onlineSavingsMsg'
Learn More

S-Ethyl thiopropionate, 97%, Thermo Scientific™

Product Code. 11451407
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451407 5 g 5g
11461407 25 g 25g
11471407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451407 Supplier Thermo Scientific Alfa Aesar Supplier No. B24918.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-42-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00027016
InChI Key HNEVHBHRLCAKKQ-UHFFFAOYSA-N
Synonym s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester
PubChem CID 75513
IUPAC Name S-ethyl propanethioate
SMILES CCC(=O)SCC

Specifications

Density 0.958
Boiling Point 137°C to 138°C
Flash Point 27°C (80°F)
Odor Stench
Refractive Index 1.459
Quantity 5 g
UN Number UN1993
Beilstein 1740740
Formula Weight 118.2
Percent Purity 97%
Chemical Name or Material S-Ethyl thiopropionate
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.