missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Glycidol, 97%, (98% ee)

Product Code. 10346884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346884 1 g Glass bottle 1g
10103444 5 g Glass bottle 5g
10224684 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10346884 Supplier Thermo Scientific Acros Supplier No. 411000010
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 60456-23-7 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60456-23-7
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00074874
InChI Key CTKINSOISVBQLD-VKHMYHEASA-N
Synonym s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s
PubChem CID 6973630
ChEBI CHEBI:38690
IUPAC Name [(2S)-oxiran-2-yl]methanol
SMILES C1C(O1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -54°C
Color Colorless to Yellow
Density 1.1160g/mL
Boiling Point 160°C to 161°C
Flash Point 66°C
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4340 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.116
Specific Rotation −12° to −18° (20°C, 589nm) (neat)
Solubility Information Solubility in water: soluble
Formula Weight 74.08
Percent Purity 97%,(98% ee)
Physical Form Liquid
Chemical Name or Material (S)-(-)-Glycidol, 98% ee
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-Glycidol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H350-May cause cancer.
  • H360F-May damage fertility.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.