Learn More
(S)-(-)-Glycidol, 99+%, ee 99+%
Description
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(S)-(-)-Glycidol is employed in the synthesis of chiral (E)-allylic alcohol side-chains for introduction into prostacyclin and prostaglandin frameworks by cross-metathesis. Also used as a starting reagent to synthesize non-peptidic HIV protease inhibitors.
Solubility
Soluble in chloroform, DMSO and methanol.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Color | Yellow |
| Density | 1.116 g/mL |
| Boiling Point | 66°C to 67°C (19 mmHg) |
| Flash Point | 66°C (151°F) |
| Refractive Index | 1.433 |
| Quantity | 25 g |
| UN Number | UN2810 |
| Beilstein | 79783 |
| Solubility Information | Soluble in chloroform,DMSO and methanol. |
| Formula Weight | 74.08 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.