missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Ibuprofen (S)-(+)-Lysinate, TRC

Product Code. 30395491
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395491 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395491 Supplier Toronto Research Chemicals Supplier No. TRCI14002525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(+)-Ibuprofen (S)-(+)-Lysinate
CAS 113403-10-4
Molecular Formula C13 H18 O2 . C6 H14 N2 O2
Quantity 25 mg
Synonym L-Lysine, (αS)-α-methyl-4-(2-methylpropyl)benzeneacetate (1:1) (ACI), L-Lysine, mono[(S)-α-methyl-4-(2-methylpropyl)benzeneacetate] (ZCI), L-Lysine, mono[(αS)-α-methyl-4-(2-methylpropyl)benzeneacetate] (9CI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (S)-, compd. with L-lysine (1:1) (ZCI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (αS)-, compd. with L-lysine (1:1) (9CI), (S)-Ibuprofen-(S)-lysine, Dexibuprofen lysinate, Dexibuprofen lysine, L 669445, L 669455, MK 233, S-(+)-Ibuprofen L-lysine salt, S-Ibuprofen lysinate
InChI Formula InChI=1S/C13H18O2.C6H14N2O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;7-4-2-1-3-5(8)6(9)10/h4-7,9-10H,8H2,1-3H3,(H,14,15);5H,1-4,7-8H2,(H,9,10)/t10-;5-/m00/s1
SMILES CC(C)Cc1ccc(cc1)[C@H](C)C(=O)O.NCCCC[C@H](N)C(=O)O
IUPAC Name (2S)-2,6-diaminohexanoic acid;(2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
Molecular Weight (g/mol) 352.47
Formula Weight 352.2362
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.