missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Indoline-2-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10525501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10525501 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10525501 Supplier Thermo Scientific Acros Supplier No. 342080050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 79815-20-6
CAS Min % 97.5
CAS Max % 100.0
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H9NO2
MDL Number MFCD00070578
Quantity 5 g
Specific Rotation Condition −112.5° (20°C c=1,1N HCl)
Specific Rotation -112.5
Synonym s---indoline-2-carboxylic acid, s-indoline-2-carboxylic acid, 2s-2,3-dihydro-1h-indole-2-carboxylic acid, h-idc-oh, l-indoline-2-carboxylic acid, l---indoline-2-carboxylic acid, s-2,3-dihydro-1h-indole-2-carboxylic acid, s---indolin-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2s
InChI Key QNRXNRGSOJZINA-QMMMGPOBSA-N
SMILES C1C(NC2=CC=CC=C21)C(=O)O
IUPAC Name (2S)-2,3-dihydro-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 163.18
PubChem CID 2733920
Formula Weight 163.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-(-)-Indoline-2-carboxylic acid, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-Indoline-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.