missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Mandelic acid, 99+%

Product Code. 11487337
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487337 5 g 5g
11497337 25 g 25g
11407347 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487337 Supplier Thermo Scientific Alfa Aesar Supplier No. A15061.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17199-29-0 | C8H8O3 | 152.149 g/mol

(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17199-29-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004495
InChI Key IWYDHOAUDWTVEP-ZETCQYMHSA-N
Synonym s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
PubChem CID 439616
ChEBI CHEBI:32800
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 131°C to 135°C
Solubility Information Soluble in water.
Formula Weight 152.15
Percent Purity ≥99%
Quantity 5 g
Optical Rotation +155° (c=5 in Water)
Sensitivity Light sensitive
Chemical Name or Material (S)-(+)-Mandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.