missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Mandelic Acid, TRC

Product Code. 30138010
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138010 25 g 25g
30004734 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138010 Supplier Toronto Research Chemicals Supplier No. TRCM16253525G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17199-29-0
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym Benzeneacetic acid, α-hydroxy-, (αS)-, Benzeneacetic acid, α-hydroxy-, (S)-, Mandelic acid, L-(+)- (8CI), (αS)-α-Hydroxybenzeneacetic acid, (+)-(S)-Mandelic acid, (+)-Mandelic acid, (+)-α-Hydroxyphenylacetic acid, (2S)-2-Hydroxy-2-(phenyl)ethanoic acid, (2S)-2-Hydroxy-2-phenylacetic acid, (S)-(+)-Hydroxyphenylacetic acid, (S)-(+)-Mandelic acid, (S)-2-Hydroxy-2-(phenyl)ethanoic acid, (S)-2-Hydroxy-2-phenylacetic acid, (S)-Amygdalic acid, (S)-Mandelic acid, (S)-α-Hydroxybenzeneacetic acid, (S)-α-Hydroxyphenylacetic acid, L-(+)-Mandelic acid, L-Mandelic acid, Atomoxetine Hydrochloride Imp E (EP)
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
SMILES O[C@H](C(=O)O)c1ccccc1

Specifications

InChI Formula InChI=1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m0/s1
Formula Weight 152.0473
Quantity 25 g
Chemical Name or Material L-(+)-Mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.