missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl methanethiolsulfonate, 97%

Product Code. 10197783 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10197783 25 g Glass bottle 25g
10171043 1 g Glass bottle 1g
10793514 5 g Glass bottle 5g
11945731 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10197783 Supplier Thermo Scientific Acros Supplier No. 197390250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2949-92-0 | C2H6O2S2 | 126.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2949-92-0
Molecular Formula C2H6O2S2
Molecular Weight (g/mol) 126.19
MDL Number MFCD00007565
InChI Key XYONNSVDNIRXKZ-UHFFFAOYSA-N
Synonym s-methyl methanethiosulfonate, methyl methanethiolsulfonate, methyl methanethiosulfonate, s-methyl methanesulfonothioate, methanesulfonothioic acid, s-methyl ester, methyl methanesulfonothioate, s-methyl methanethiolsulfonate, methanethiosulfonic acid s-methyl ester, s-methyl methanethiosulphonate, methanesulfonylsulfanyl methane
PubChem CID 18064
ChEBI CHEBI:74357
IUPAC Name methylsulfonylsulfanylmethane
SMILES CSS(=O)(=O)C

Specifications

Density 1.2270g/mL
Boiling Point 69.0°C to 71.0°C (0.4 mmHg)
Flash Point 87°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3SO2SCH3
Refractive Index 1.5118
Quantity 25 g
Fieser 05,454; 07,243; 12,325; 01,169; 10,107; 16,99
Specific Gravity 1.227
Formula Weight 126.19
Percent Purity 97%
Chemical Name or Material S-Methyl methanethiolsulfonate
Show More Show Less
compliance-icons
Product Identifier
  • S-Methyl methanethiolsulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.