missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl-N,N-diethylthiocarbamate Sulfoxide, TRC

Product Code. 30010301
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010301 5 mg 5mg
30010319 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010301 Supplier Toronto Research Chemicals Supplier No. TRCM3017805MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140703-15-7
Molecular Formula C6 H13 N O2 S
Molecular Weight (g/mol) 163.24
Synonym Disulfiram Impurity 4
IUPAC Name N,N-diethyl-1-methylsulfinylformamide
SMILES CCN(CC)C(=O)S(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H13NO2S/c1-4-7(5-2)6(8)10(3)9/h4-5H2,1-3H3
Formula Weight 163.0667
Chemical Name or Material S-Methyl-N,N-diethylthiocarbamate Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.