missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl thiobutyrate, 98%

Product Code. 11439667
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439667 25 g 25g
11429667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439667 Supplier Thermo Scientific Alfa Aesar Supplier No. B22144.14
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2432-51-1 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-51-1
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00009872
InChI Key GRLJIIJNZJVMGP-UHFFFAOYSA-N
Synonym s-methyl thiobutyrate, butanethioic acid, s-methyl ester, methyl thiolbutyrate, methanethiol butyrate, methanethiol n-butyrate, butyric acid, thio-, s-methyl ester, s-methyl thiobutanoate, unii-2p1e432myz, methyl thiobutanoate, methyl thiobutyrate natural
PubChem CID 62444
IUPAC Name S-methyl butanethioate
SMILES CCCC(=O)SC

Specifications

Density 0.966
Boiling Point 142°C to 143°C
Flash Point 34°C (93°F)
Refractive Index 1.462
Quantity 25 g
UN Number UN1993
Beilstein 1848987
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material S-Methyl thiobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.