missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies (S)-MG132

Product Code. 17180192
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17180192 10 mg 10mg
17170192 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17180192 Supplier STEMCELL Technologies Supplier No. 73264
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ubiquitin-proteasome and NF-κB pathway inhibitor; Inhibits proteasome and stabilizes IκB

  • (S)-MG132 is a reversible, cell-permeable inhibitor of proteasome activity (IC50 = 100 nM; Kisselev & Goldberg) and calpain (IC50 = 1.2 μM; Tsubuki et al.) The ubiquitin-proteasome pathway selectively degrades intracellular proteins, thereby clearing damaged or misfolded proteins, and regulating the availability of key proteins involved in the control of inflammatory processes and cell cycle regulation
  • (S)-MG132 suppresses NF-κB activation by preventing IκB degradation (IC50 = 3 μM; Arlt et al.), Palombella et al.), Ortiz-Lazareno et al.)
  • Blocks apoptosis triggered by DNA damage in HeLa cells (Zhang et al.)
  • Inhibits NF-κB activation, sensitizing a variety of carcinoma cell lines to apoptosis (Arlt et al.)
  • Cytotoxic effects on a variety of human cancer cell lines (Banerjee & Liefshitz)
  • Inhibits growth of mouse melanoma (B16) and human ocular melanoma (IPC227F) cell lines (Vivier et al.)

Specifications

CAS 133407-82-6
Synonym Z-Leu-Leu-Leu-CHO; Calpain inhibitor IV-2
Quantity 10 mg
Molecular Formula C26H41N3O5
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.