missing translation for 'onlineSavingsMsg'
Learn More

S-Mirtazapine, TRC

Product Code. 30483952
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483952 10 mg 10mg
30483554 25 mg 25mg
30480886 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483952 Supplier Toronto Research Chemicals Supplier No. TRCM36498510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61337-87-9
Molecular Formula C17 H19 N3
Molecular Weight (g/mol) 265.35
Synonym Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, (14bS)-, Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, (S)-, (14bS)-1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, (+)-Mirtazapine, (S)-6-Azamianserin, (S)-Mirtazapine, (S)-Org 3770, Esmirtazapine, Org 44-20, S-(+)-Mirtazapine
SMILES CN1CCN2[C@H](C1)c3ccccc3Cc4cccnc24

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3/t16-/m1/s1
Formula Weight 265.1579
Chemical Name or Material (S)-Mirtazapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.