missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Despropyl Pramipexole, TRC

Product Code. 30138568
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138568 1 g 1g
30134230 10 g 10g
30007594 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30138568 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD4168401G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106092-09-5
Molecular Formula C7 H11 N3 S
Molecular Weight (g/mol) 169.25
Synonym (S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole, (6S)-4,5,6,7-Tetrahydro-1,3-benzothiazole-2,6-diamine, (S)-N-Despropyl Pramipexole
IUPAC Name (6S)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
SMILES N[C@H]1CCc2nc(N)sc2C1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H11N3S/c8-4-1-2-5-6(3-4)11-7(9)10-5/h4H,1-3,8H2,(H2,9,10)/t4-/m0/s1
Formula Weight 169.0674
Chemical Name or Material (6S)-4,5,6,7-Tetrahydro-1,3-benzothiazole-2,6-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.