missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Despropyl Pramipexole, TRC

Product Code. 30007594
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007594 100 mg 100mg
30138568 1 g 1g
30134230 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007594 Supplier Toronto Research Chemicals Supplier No. TRCD416840100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106092-09-5
Formula Weight 169.0674
InChI Formula InChI=1S/C7H11N3S/c8-4-1-2-5-6(3-4)11-7(9)10-5/h4H,1-3,8H2,(H2,9,10)/t4-/m0/s1
IUPAC Name (6S)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
Molecular Formula C7 H11 N3 S
Molecular Weight (g/mol) 169.25
SMILES N[C@H]1CCc2nc(N)sc2C1
Synonym (S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole, (6S)-4,5,6,7-Tetrahydro-1,3-benzothiazole-2,6-diamine, (S)-N-Despropyl Pramipexole
Chemical Name or Material (6S)-4,5,6,7-Tetrahydro-1,3-benzothiazole-2,6-diamine

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.