missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N alpha-Dimethylbenzylamine, 97%

Product Code. 11995241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995241 1 g Glass bottle 1g
11905251 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995241 Supplier Thermo Scientific Acros Supplier No. 395840010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19131-99-8 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19131-99-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key RCSSHZGQHHEHPZ-QMMMGPOBSA-N
Synonym s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
PubChem CID 2060073
IUPAC Name (1S)-N-methyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NC

Specifications

Density 0.9240g/mL
Boiling Point 74.0°C to 76.0°C (11.0 mmHg)
Flash Point 61°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NHCH3
Quantity 1 g
Specific Gravity 0.924
Solubility Information Solubility in water: insoluble
Formula Weight 135.21
Percent Purity 99+%
Chemical Name or Material (S)-(-)-N α-Dimethylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-N alpha-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: (S)-(-)-N alpha-Dimethylbenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.