missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Fmoc-N6,N6-dimethyl-L-lysine Hydrochloride, TRC

Product Code. 30408271
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408271 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408271 Supplier Toronto Research Chemicals Supplier No. TRCF648400500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-N-Fmoc-N6,N6-dimethyl-L-lysine hydrochloride
CAS 252049-10-8
Molecular Formula C23 H28 N2 O4 . Cl H
Quantity 500 mg
Synonym N2-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N6,N6-dimethyl-L-Lysine Hydrochloride, (S)-N-Fmoc-N',N''-dimethyl-L-lysine Hydrochloride
InChI Formula InChI=1S/C23H28N2O4.ClH/c1-25(2)14-8-7-13-21(22(26)27)24-23(28)29-15-20-18-11-5-3-9-16(18)17-10-4-6-12-19(17)20;/h3-6,9-12,20-21H,7-8,13-15H2,1-2H3,(H,24,28)(H,26,27);1H/t21-;/m0./s1
SMILES Cl.CN(C)CCCC[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC Name (2S)-6-(dimethylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid;hydrochloride
Molecular Weight (g/mol) 432.94
Formula Weight 432.1816
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.