missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine, tech. 85%

Product Code. 11368716
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368716 5 g 5g
11358716 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368716 Supplier Alfa Aesar™ Supplier No. L19414.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 143140-08-3
Molecular Formula C14H25NOSi
Molecular Weight (g/mol) 251.445
MDL Number MFCD06798960
InChI Key FRQGYHCIEIDEAD-ZDUSSCGKSA-N
Synonym s-n-methoxymethyl-1-phenyl-n-trimethylsilyl methyl ethanamine, methoxymethyl 1s-1-phenylethyl trimethylsilyl methyl amine, s---n-methoxymethyl-n-trimethylsilyl methyl-1-phenylethylamine, s---n-methoxymethyl-n-trimethylsilyl methyl-1, s-methoxymethyl-1-phenyl-ethyl-trimethylsilanylmethyl-amine, s-n-methoxymethyl-n-trimethylsilyl methyl-1-phenylethylamine, s---n-methoxymethyl-n-trimethylsilylmethyl-1-phenylethylamine, 1s-n-methoxymethyl-1-phenyl-n-trimethylsilyl methyl ethan-1-amine, s-+-n-methoxymethyl-n-trimethylsilyl methyl-1-phenylethylamine, s-n-methoxymethyl-1-phenyl-n-trimethylsilyl methyl ethan-1-amine
PubChem CID 10729457
IUPAC Name (1S)-N-(methoxymethyl)-1-phenyl-N-(trimethylsilylmethyl)ethanamine
SMILES CC(C1=CC=CC=C1)N(COC)C[Si](C)(C)C

Specifications

Boiling Point 82°C to 84°C (0.2 mmHg)
Quantity 5 g
Beilstein 5906612
Sensitivity Moisture sensitive
Optical Rotation −47° (c=1 in Carbon Tetrachloride)
Formula Weight 251.45
Percent Purity 85%
Grade Technical
Chemical Name or Material (S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.