missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-O-Acetylmandelic acid, 99%

Product Code. 15463068
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463068 5 g 5g
15453068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463068 Supplier Thermo Scientific Alfa Aesar Supplier No. H56567.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7322-88-5 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7322-88-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00064215,MFCD00662355,MFCD00004249
InChI Key OBCUSTCTKLTMBX-UHFFFAOYNA-N
Synonym s-2-acetoxy-2-phenylacetic acid, +-o-acetyl-l-mandelic acid, s-+-alpha-acetoxyphenylacetic acid, s-acetylmandelic acid, 2s-2-acetyloxy-2-phenylacetic acid, s-o-acetylmandelic acid, mandelic acid, acetate, l, unii-0n6kt18k98, l-+-o-acetylmandelic acid, s-+-o-acetyl-l-mandelic acid
PubChem CID 6923379
SMILES CC(=O)OC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 97°C to 100°C
Quantity 5 g
Beilstein 2694109
Optical Rotation +153° (c=2 in Acetone)
IUPAC Name (2S)-2-acetyloxy-2-phenylacetic acid
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material (S)-(+)-O-Acetylmandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.