missing translation for 'onlineSavingsMsg'
Learn More

(S)-O-Acetylmandelic Acid, TRC

Product Code. 30309799
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309799 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309799 Supplier Toronto Research Chemicals Supplier No. TRCA185960500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-O-Acetylmandelic Acid
CAS 7322-88-5
Molecular Formula C10H10O4
Quantity 500 mg
Synonym (alphaS)-alpha-(Acetyloxy)benzeneacetic Acid, L-Mandelic Acid Acetate, (2S)-2-Acetyloxy-2-phenylacetic Acid, (S)-(+)-O-Acetyl-L-mandelic Acid, (S)-2-Acetoxy-2-phenylacetic Acid, (S)-O-Acetylmandelic Acid, L-(+)-O-Acetylmandelic Acid, L-O-Acetylmandelic Acid, S-Acetylmandelic Acid
InChI Formula InChI=1S/C10H10O4/c1-7(11)14-9(10(12)13)8-5-3-2-4-6-8/h2-6,9H,1H3,(H,12,13)/t9-/m0/s1
SMILES CC(=O)O[C@H](C(=O)O)c1ccccc1
IUPAC Name (2S)-2-acetyloxy-2-phenylacetic acid
Molecular Weight (g/mol) 194.18
Formula Weight 194.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.