missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Pantoprazole Sodium Salt, TRC

Product Code. 30420969
Change view
Click to view available options
Quantity:
1 mg
5 mg
100 mg
Unit Size:
100mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420969 100 mg 100mg
30414335 5 mg 5mg
30414769 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420969 Supplier Toronto Research Chemicals Supplier No. TRCP182995100MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160488-53-9
Molecular Formula C16 H14 F2 N3 O4 S . Na
Molecular Weight (g/mol) 405.35
Synonym 1H-Benzimidazole, 6-(difluoromethoxy)-2-[(S)-[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-, sodium salt (1:1), 1H-Benzimidazole, 5-(difluoromethoxy)-2-[(S)-[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-, sodium salt (9CI), (S)-(-)-Pantoprazole sodium salt
IUPAC Name sodium;5-(difluoromethoxy)-2-[(S)-(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide
SMILES [Na+].COc1ccnc(C[S@](=O)c2nc3cc(OC(F)F)ccc3[n-]2)c1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H14F2N3O4S.Na/c1-23-13-5-6-19-12(14(13)24-2)8-26(22)16-20-10-4-3-9(25-15(17)18)7-11(10)21-16;/h3-7,15H,8H2,1-2H3;/q-1;+1/t26-;/m0./s1
Formula Weight 405.0571
Chemical Name or Material (S)-Pantoprazole sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.