missing translation for 'onlineSavingsMsg'
Learn More

(S)-Perillaldehyde, TRC

Product Code. 30386492
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386492 100 mg -
30377005 500 mg 500mg
30371300 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386492 Supplier Toronto Research Chemicals Supplier No. TRCE560003100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18031-40-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.21
Synonym (S)-4-(Prop-1-En-2-yl)cyclohex-1-enecarbaldehyde
IUPAC Name (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
SMILES CC(=C)[C@H]1CCC(=CC1)C=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H14O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,7,10H,1,4-6H2,2H3/t10-/m1/s1
Formula Weight 150.1045
Chemical Name or Material (S)-Perillaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.