missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (S)-PFI 2 hydrochloride

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
(S)-PFI 2 hydrochloride is a negative control of (R)-PFI 2 hydrochloride . Exhibits 500-fold lower activity in a SETD7 enzymatic assay (IC50 = 1 μM) compared to the active enantiomer.
Active Analog also available.
Specifications
Specifications
| CAS | 1627607-88-8 |
| Quantity | 10 mg |
| Molecular Formula | 8-Fluoro-1,2,3,4-tetrahydro-N-[(1 S)-2-oxo-2-(1-pyrrolidinyl)-1-[[3-(trifluoromethyl)phenyl]methyl]ethyl]-6-isoquinolinesulfonamide hydrochloride |
| Product Type | (S)-PFI 2 hydrochloride |
| Purity | 97% |
| Molecular Weight (g/mol) | M.W. (Observed): 535.98 |
| Inhibitors | (S)-PFI 2 hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction