missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Prolinol, 98%

Product Code. 11322267
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322267 1 g 1g
11332267 5 g 5g
11342267 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322267 Supplier Thermo Scientific Alfa Aesar Supplier No. L09779.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23356-96-9 | C5H11NO | 101.149 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water. Soluble in chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23356-96-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00005255
InChI Key HVVNJUAVDAZWCB-YFKPBYRVSA-N
Synonym l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
PubChem CID 640091
IUPAC Name [(2S)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Refractive Index 1.4853
Density 1.025
Boiling Point 74°C to 76°C (2mmHg)
Melting Point 42°C to 44°C
Quantity 1 g
Flash Point 86°C (186°F)
Optical Rotation +31° (c=1 in Toluene)
Sensitivity Air sensitive; Hygroscopic
Beilstein 79843
Solubility Information Fully miscible in water. Soluble in chloroform.
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material (S)-(+)-Prolinol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-Prolinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: L-(Pyrrolidin-1-yl)methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.