missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress (S,R,S)-AHPC-PEG2-N3

Product Code. p-200104391
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
This item is not returnable. View return policy

Product Code. 16490086

Brand: MedChemExpress HY103599

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-PEG2-N3 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 2-unit PEG linker used in PROTAC technology.

Specifications

Molecular Weight (g/mol) 601.72 g/mol
CAS 2010159-45-0
Content And Storage -20°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen)
Shipping Condition Room temperature
Quantity 2 g
Synonym VH032-PEG2-N3 ∣ VHL Ligand-Linker Conjugates 6 ∣ E3 ligase Ligand-Linker Conjugates 13
Solubility Ethanol : 100 mg/mL (166.19 mM; Need ultrasonic) ∣ DMSO : 50 mg/mL (83.10 mM; Need ultrasonic)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.