missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine, 97+%

Product Code. 15445956
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15445956

Brand: Alfa Aesar™ H60702.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1091606-67-5
Molecular Formula C26H24NP
Molecular Weight (g/mol) 381.46
MDL Number MFCD11045444
InChI Key MIPGTOGPLXZBQX-QYRYXDJWNA-N
Synonym 1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
PubChem CID 46177587
IUPAC Name [(1S,2S)-2-amino-1,2-diphenylethyl]diphenylphosphane
SMILES N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C to 93°C
Quantity 250 mg
Optical Rotation +179° (c=0.5 in Chloroform)
Formula Weight 318.45
Percent Purity ≥97%
Chemical Name or Material (S,S)-(+)-2-Diphenylphosphino-1,2-diphenylethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.